Benzyl cinnamate 98%

Supplier: Ambeed
Warning

A377657-100G A377657-250MG A377657-25G A377657-500G
76694-420EA 14 USD
76694-420 76694-422 76694-424 76694-426
Benzyl cinnamate 98%
Benzyl cinnamate

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₂
MDL Number: MFCD00004789
CAS Number: 103-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR