N-(3-Cyano-7-ethoxy-4-hydroxyquinolin-6-yl)acetamide 95%

Supplier: Ambeed
Warning

A377546-100MG A377546-250MG
76694-410EA 13.8 USD
76694-410 76694-412
N-(3-Cyano-7-ethoxy-4-hydroxyquinolin-6-yl)acetamide 95%
N-(3-Cyano-7-ethoxy-4-hydroxyquinolin-6-yl)acetamide

Precaución: for Research Use Only

Formula: C₁₄H₁₃N₃O₃
MW: 271.27 g/mol
MDL Number: MFCD09029047
CAS Number: 848133-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR