ɑ,ɑ,ɑ'-Tris(4-hydroxyphenyl)-1-ethyl-4-isopropylbenzene 98%

Supplier: Ambeed
Warning

Synonyms: 4-[4-[1,1-Bis(4-hydroxyphenyl)ethyl]]-α,α-dimethylbenzylphenol, 4,4'-(1-(4-(2-(4-Hydroxyphenyl)propan-2-yl)phenyl)ethane-1,1-diyl)diphenol, α,α,α'-Tris(4-hydroxyphenyl)-1-ethyl-4-isopropylbenzene, 4-[4-[1,1-Bis(4-hydroxyphenyl)ethyl]]-ɑ,ɑ-dimethylbenzylphenol

A377030-10G A377030-1G A377030-5G
76694-404EA 23.34 USD
76694-404 76694-406 76694-408
ɑ,ɑ,ɑ'-Tris(4-hydroxyphenyl)-1-ethyl-4-isopropylbenzene 98%
alpha,alpha,alpha'-Tris(4-hydroxyphenyl)-1-ethyl-4-isopropylbenzene

Precaución: for Research Use Only

Formula: C₂₉H₂₈O₃
MW: 424.54 g/mol
MDL Number: MFCD00191685
CAS Number: 110726-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR