Benzo[b]thiophene-2-methanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Hydroxymethyl)benzo[b]thiophene, Benzo[b]thiophen-2-ylmethanol

A376920-10G A376920-1G A376920-250MG A376920-25G A376920-5G A376920-100G
76694-392EA 40 USD
76694-392 76694-394 76694-396 76694-398 77594-130 77878-095
Benzo[b]thiophene-2-methanol 97%
Benzo[b]thiophene-2-methanol

Precaución: for Research Use Only

Formula: C₉H₈OS
MDL Number: MFCD02682004
CAS Number: 17890-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR