Biphenylene 95%

Supplier: Ambeed
Warning

A376298-100MG A376298-1G A376298-250MG
76694-384EA 92.3 USD
76694-384 76694-386 76694-388
Biphenylene 95%
Biphenylene

Precaución: for Research Use Only

Formula: C₁₂H₈
MW: 152.2 g/mol
MDL Number: MFCD00001110
CAS Number: 259-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR