Benzydamine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A374095-100G
A374095-5G
76694-368EA
109
USD
76694-368
76694-370
76694-372
76694-374
Benzydamine hydrochloride 98%
Benzydamine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₉H₂₃N₃O |
MDL Number:
MFCD00078957 CAS Number: 132-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...