(1R,5S)-endo-8-Benzyl-8-azabicyclo[3.2.1]octan-3-amine 97%

Supplier: Ambeed
Warning

A373718-100MG A373718-1G A373718-250MG A373718-5G
76694-360EA 10 USD
76694-360 76694-362 76694-364 76694-366
(1R,5S)-endo-8-Benzyl-8-azabicyclo[3.2.1]octan-3-amine 97%
(1R,5S)-endo-8-Benzyl-8-azabicyclo[3.2.1]octan-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂
MDL Number: MFCD08460051
CAS Number: 76272-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR