8-Chlorooct-1-ene 97%

Supplier: Ambeed
Danger

Synonyms: Oct-7-en-1-yl chloride

A372091-1G A372091-250MG A372091-5G
76694-352EA 46.68 USD
76694-352 76694-354 76694-356
8-Chlorooct-1-ene 97%
8-Chlorooct-1-ene

Precaución: for Research Use Only

Formula: C₈H₁₅Cl
MW: 146.658 g/mol
MDL Number: MFCD00671827
CAS Number: 871-90-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR