Calcium hydrogen orthophosphate dihydrate 99%

Supplier: Ambeed
Warning

Synonyms: Calcium phosphate dibasic dihydrate, Calcium hydrogen phosphate dihydrate, Calcium hydrogenphosphate dihydrate, Dibasic Calcium Phosphate Dihydrate

A371796-1000G A371796-100G A371796-500G
76694-346EA 22 USD
76694-346 76694-348 76694-350
Calcium hydrogen orthophosphate dihydrate 99%
Calcium hydrogen orthophosphate dihydrate

Precaución: for Research Use Only

Formula: HCaO₄P·2H₂O
MDL Number: MFCD00149621
CAS Number: 7789-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR