(S)-(((1-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3-hydroxypropan-2-yl)oxy)methyl)phosphonic acid 99%

Supplier: Ambeed
Danger

Synonyms: Cidofovir

A370831-1MG A370831-100MG A370831-250MG
76694-332EA 5.31 USD
76694-332 77551-072 77626-718
(S)-(((1-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3-hydroxypropan-2-yl)oxy)methyl)phosphonic acid 99%
(S)-(((1-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3-hydroxypropan-2-yl)oxy)methyl)phosphonic acid

Precaución: for Research Use Only

Formula: C₈H₁₄N₃O₆P
MW: 279.19 g/mol
MDL Number: MFCD00866936
CAS Number: 113852-37-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR