Isopropyl carbamate 98%

Supplier: Ambeed
Warning

Synonyms: Carbamic Acid Isopropyl Ester

A370806-100G A370806-25G A370806-5G
76694-328EA 150 USD
76694-328 76694-330 77878-087
Isopropyl carbamate 98%
Isopropyl carbamate

Precaución: for Research Use Only

Formula: C₄H₉NO₂
MDL Number: MFCD00025463
CAS Number: 1746-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR