Bromohydroquinone 90%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromobenzene-1,4-diol
A370443-1G
A370443-250MG
A370443-25G
A370443-5G
76694-320EA
40.32
USD
76694-320
76694-322
76694-324
76694-326
Bromohydroquinone 90%
Bromohydroquinone
Precaución: for Research Use Only
Formula:
C₆H₅BrO₂ MW: 189.01 g/mol |
MDL Number:
MFCD00041747 CAS Number: 583-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...