Biurea 95%

Supplier: Ambeed
Warning

A370043-100G A370043-1G A370043-5G
76694-314EA 327 USD
76694-314 76694-316 76694-318
Biurea 95%
Biurea

Precaución: for Research Use Only

Formula: C₂H₆N₄O₂
MDL Number: MFCD00025398
CAS Number: 110-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR