(1S,2S)-2-Amino-1-(4-aminophenyl)propane-1,3-diol 98%

Supplier: Ambeed
Warning

A369883-100MG A369883-1G A369883-250MG
76694-304EA 13.8 USD
76694-304 76694-306 76694-308
(1S,2S)-2-Amino-1-(4-aminophenyl)propane-1,3-diol 98%
(1S,2S)-2-Amino-1-(4-aminophenyl)propane-1,3-diol

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂
MW: 182.21 g/mol
MDL Number: MFCD00238534
CAS Number: 50961-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR