7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine 95%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-4-methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine, 7-Bromo-3,4-dihydro-4-methyl-2H-1,4-benzoxazine

A369541-250MG A369541-5G A369541-100MG
76694-300EA 72.15 USD
76694-300 76694-302 77601-634
7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine 95%
7-Bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MW: 228.1 g/mol
MDL Number: MFCD02681913
CAS Number: 154264-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR