Methyl acetylsalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-acetoxybenzoate, Methyl 2-Acetoxybenzoate, Aspirin methyl ester, Acetylsalicylic Acid Methyl Ester, 2-Acetoxybenzoic Acid Methyl Ester

A369424-25G A369424-100G A369424-500G
76694-298EA 28 USD
76694-298 77601-632 77626-712
Methyl acetylsalicylate 98%
Methyl acetylsalicylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00014978
CAS Number: 580-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR