Benzyl methyl malonate 98%

Supplier: Ambeed
Warning

A368064-1G A368064-5G
76694-288EA 8.49 USD
76694-288 76694-290
Benzyl methyl malonate 98%
Benzyl methyl malonate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD00008461
CAS Number: 52267-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR