3-Hydroxycinnamic acid 95+%

Supplier: Ambeed
Warning

Synonyms: m-Hydroxycinnamic acid, 3-(3-Hydroxyphenyl)acrylic acid, m-Coumaric acid

A367108-100G A367108-25G A367108-5G
76694-282EA 201.58 USD
76694-282 76694-284 76694-286
3-Hydroxycinnamic acid 95+%
3-Hydroxycinnamic acid

Precaución: for Research Use Only

Formula: C₉H₈O₃
MW: 164.158 g/mol
MDL Number: MFCD00004390
CAS Number: 588-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR