1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Bromoethoxy)benzotrifluoride

A366981-1G A366981-5G
76694-278EA 33.95 USD
76694-278 76694-280
1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene 98%
1-(2-Bromoethoxy)-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈BrF₃O
MW: 269.06 g/mol
MDL Number: MFCD00215501
CAS Number: 18800-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR