Chlorohydridotris(triphenylphosphine)ruthenium(II) toluene adduct 98%

Supplier: Ambeed
Warning

Synonyms: Tris(triphenylphosphoranyl)ruthenium(V) chloride hydride

A366869-100MG A366869-1G A366869-250MG A366869-25G A366869-5G
76694-268EA 33.95 USD
76694-268 76694-270 76694-272 76694-274 76694-276
Chlorohydridotris(triphenylphosphine)ruthenium(II) toluene adduct 98%
Chlorohydridotris(triphenylphosphine)ruthenium(II) toluene adduct

Precaución: for Research Use Only

Formula: C₅₄H₄₉ClP₃Ru
MW: 927.4112 g/mol
MDL Number: MFCD04971968
CAS Number: 55102-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR