Chloromethyldimethylethoxysilane 97%

Supplier: Ambeed
Danger

Synonyms: (Chloromethyl)(ethoxy)dimethylsilane

A365316-1G A365316-25G A365316-5G
76694-262EA 7.43 USD
76694-262 76694-264 76694-266
Chloromethyldimethylethoxysilane 97%
Chloromethyldimethylethoxysilane

Precaución: for Research Use Only

Formula: C₅H₁₃ClOSi
MW: 152.695 g/mol
MDL Number: MFCD00045263
CAS Number: 13508-53-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR