1,2-O-Isopropylidene-ɑ-D-glucurono-6,3-lactone 98%

Supplier: Ambeed
Warning

Synonyms: (3aR,3bS,6S,6aR,7aR)-6-Hydroxy-2,2-dimethyltetrahydrofuro[2',3':4,5]furo[2,3-d][1,3]dioxol-5(3aH)-one

A365252-100G A365252-1G A365252-25G A365252-500G A365252-5G
76694-252EA 129.43 USD
76694-252 76694-254 76694-256 76694-258 76694-260
1,2-O-Isopropylidene-ɑ-D-glucurono-6,3-lactone 98%
1,2-O-Isopropylidene-alpha-D-glucurono-6,3-lactone

Precaución: for Research Use Only

Formula: C₉H₁₂O₆
MW: 216.19 g/mol
MDL Number: MFCD00061641
CAS Number: 20513-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR