Diallyldiphenylsilane 95%

Supplier: Ambeed
Warning

A364673-250MG A364673-25G A364673-5G A364673-1G
76694-246EA 18 USD
76694-246 76694-248 76694-250 77626-704
Diallyldiphenylsilane 95%
Diallyldiphenylsilane

Precaución: for Research Use Only

Formula: C₁₈H₂₀Si
MW: 264.44 g/mol
MDL Number: MFCD00026068
CAS Number: 10519-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR