2-Amino-2-(2,6-difluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A364609-100MG A364609-1G A364609-250MG
76694-240EA 20 USD
76694-240 76694-242 76694-244
2-Amino-2-(2,6-difluorophenyl)acetic acid 98%
2-Amino-2-(2,6-difluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₂
MDL Number: MFCD07786876
CAS Number: 244187-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR