7-Hydroxy-5-methyl-2-methylthio[1,2,4]triazolo[1,5-a]pyrimidine 98%

Supplier: Ambeed
Warning

A364112-1G A364112-5G
76694-236EA 17 USD
76694-236 76694-238
7-Hydroxy-5-methyl-2-methylthio[1,2,4]triazolo[1,5-a]pyrimidine 98%
7-Hydroxy-5-methyl-2-methylthio[1,2,4]triazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₈N₄OS
MDL Number: MFCD00186154
CAS Number: 40775-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR