Sulbutiamine 98%

Supplier: Ambeed
Warning

A363144-100G A363144-1G A363144-25G A363144-5G
76694-228EA 135 USD
76694-228 76694-230 76694-232 76694-234
Sulbutiamine 98%
Sulbutiamine

Precaución: for Research Use Only

Formula: C₃₂H₄₆N₈O₆S₂
MDL Number: MFCD01726427
CAS Number: 3286-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR