Auranofin 98%

Supplier: Ambeed
Danger

Synonyms: ((2S,3R,4S,5R,6R)-3,4,5-Triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-thio)(triethylphosphine)gold

A361974-10MG A361974-1G A361974-1MG A361974-250MG A361974-50MG A361974-100MG
76694-210EA 62.6 USD
76694-210 76694-212 76694-214 76694-216 76694-218 77626-700
Auranofin 98%
Auranofin

Precaución: for Research Use Only

Formula: C₂₀H₃₆AuO₉PS
MW: 680.4998 g/mol
MDL Number: MFCD00080759
CAS Number: 34031-32-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR