2-(4-Fluorophenyl)pyrrolidine 98%

Supplier: Ambeed
Warning

A360319-25G A360319-5G
76694-200EA 675.8 USD
76694-200 76694-202
2-(4-Fluorophenyl)pyrrolidine 98%
2-(4-Fluorophenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₂FN
MW: 165.211 g/mol
MDL Number: MFCD02663411
CAS Number: 72216-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR