Azo rubine 98% (by HPLC)

Supplier: Ambeed
Warning

A358747-1000G A358747-100G A358747-25G A358747-500G
76694-190EA 270 USD
76694-190 76694-192 76694-194 76694-196
Azo rubine 98% (by HPLC)
Azo rubine

Precaución: for Research Use Only

Formula: C₂₀H₁₄N₂O₇S₂
MDL Number: MFCD00003978
CAS Number: 3567-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR