(±)-6,6'-Dibromo-1,1'-bi-2-naphthol 98%
Supplier: Ambeed
|
Warning
|
A357031-1G
A357031-250MG
A357031-5G
76694-156EA
23
USD
76694-156
76694-158
76694-160
(±)-6,6'-Dibromo-1,1'-bi-2-naphthol 98%
(±)-6,6'-Dibromo-1,1'-bi-2-naphthol
Precaución: for Research Use Only
Formula:
C₂₀H₁₂Br₂O₂ |
MDL Number:
MFCD00798290 CAS Number: 13185-00-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...