Tranylcypromine hemisulfate 98%

Supplier: Ambeed
Danger

Synonyms: SKF trans-355, 2-PCPA, trans-2-Phenylcyclopropanamine hemisulphate

A354034-1MG A354034-25MG
76694-118EA 5.31 USD
76694-118 76694-120
Tranylcypromine hemisulfate 98%
Tranylcypromine hemisulfate

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₂O₄S
MW: 364.46 g/mol
MDL Number: MFCD00079222
CAS Number: 13492-01-8
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR