Calcium hydrogen orthophosphate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic
A351670-1000G
A351670-100G
A351670-25G
A351670-500G
76694-096EA
23.34
USD
76694-096
76694-098
76694-100
76694-102
Calcium hydrogen orthophosphate 99%
Calcium hydrogen orthophosphate
Precaución: for Research Use Only
Formula:
CaHPO₄ MW: 136.06 g/mol |
MDL Number:
MFCD00010909 CAS Number: 7757-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...