1-Cetylpyridinium chloride monohydrate 95%

Supplier: Ambeed
Danger

Synonyms: Hexadecylpyridinium chloride monohydrate, 1-Hexadecylpyridinium chloride monohydrate, Cetylpyridinium Chloride, Monohydrate, Cetylpyridinium chloride monohydrate

A349156-100MG A349156-25MG A349156-1MG
76694-084EA 6.37 USD
76694-084 76694-086 77626-664
1-Cetylpyridinium chloride monohydrate 95%
1-Cetylpyridinium chloride monohydrate

Precaución: for Research Use Only

Formula: C₂₁H₃₈ClN·1H₂O
MW: 358.01 g/mol
MDL Number: MFCD00149977
CAS Number: 6004-24-6
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR