2'-Fluoro-3'-(trifluoromethyl)acetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Fluoro-3-(trifluoromethyl)phenyl)ethanone

A348878-10G A348878-1G A348878-250MG A348878-25G A348878-5G
76694-074EA 92.3 USD
76694-074 76694-076 76694-078 76694-080 76694-082
2'-Fluoro-3'-(trifluoromethyl)acetophenone 97%
2'-Fluoro-3'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₆F₄O
MW: 206.14 g/mol
MDL Number: MFCD00061145
CAS Number: 207853-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR