4-Methoxybenzyl isocyanate 95%

Supplier: Ambeed
Danger

A347523-100MG A347523-1G A347523-250MG A347523-5G
76694-060EA 15 USD
76694-060 76694-062 76694-064 76694-066
4-Methoxybenzyl isocyanate 95%
4-Methoxybenzyl isocyanate

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MDL Number: MFCD00673064
CAS Number: 56651-60-6
UN: 2206
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR