Difurfuryl sulfide 98%

Supplier: Ambeed
Warning

Synonyms: Bis(furan-2-ylmethyl)sulphane

A347072-25G A347072-5G A347072-1G
76694-056EA 20 USD
76694-056 76694-058 77878-055
Difurfuryl sulfide 98%
Difurfuryl sulphide

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂S
MW: 194.2500 g/mol
MDL Number: MFCD00010081
CAS Number: 13678-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR