H-β-HoPhe-OH.HCl 95%

Supplier: Ambeed
Warning

Synonyms: L-β-Homophenylalanine hydrochloride, (S)-3-Amino-4-phenylbutyric acid hydrochloride

A345356-250MG A345356-1G
76694-042EA 36.08 USD
76694-042 77626-662
H-β-HoPhe-OH.HCl 95%
H-β-HoPhe-OH.HCl

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MW: 215.67 g/mol
MDL Number: MFCD01862875
CAS Number: 138165-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR