4'-Iodoacetophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Iodophenyl)ethan-1-one, p-Iodoacetophenone, 1-(4-Iodophenyl)ethanone
A344622-1000G
A344622-100G
A344622-10G
A344622-500G
A344622-5G
A344622-25G
76694-032EA
387
USD
76694-032
76694-034
76694-036
76694-038
76694-040
76839-944
4'-Iodoacetophenone 98%
4'-Iodoacetophenone
Precaución: for Research Use Only
Formula:
C₈H₇IO |
MDL Number:
MFCD00045320 CAS Number: 13329-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...