Cyclothiazide 98+% (mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-3,4-dihydro-3-(2-norbornen-5-yl)-2H-1,2-4-benzothiadiazine-7-sulphonamide 1,1-Dioxide, Cyclothiazide (mixture of isomers)

A343078-100MG A343078-1MG
76693-994EA 191 USD
76693-994 76693-996
Cyclothiazide 98+% (mixture of isomers)
Cyclothiazide

Precaución: for Research Use Only

Formula: C₁₄H₁₆ClN₃O₄S₂
MDL Number: MFCD00210192
CAS Number: 2259-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR