2-Methoxyphenylacetone 98%

Supplier: Ambeed
Warning

A342643-1G A342643-250MG
76693-990EA 22 USD
76693-990 76693-992
2-Methoxyphenylacetone 98%
2-Methoxyphenylacetone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MDL Number: MFCD00008770
CAS Number: 5211-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR