Bis(4-formylphenyl)phenylamine 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(Phenylimino)dibenzaldehyde, N,N-Bis(4-formylphenyl)aniline, 4,4'-(Phenylazanediyl)dibenzaldehyde, 4,4'-Diformyltriphenylamine

A342314-100MG A342314-250MG A342314-5G
76693-984EA 31.83 USD
76693-984 76693-986 76693-988
Bis(4-formylphenyl)phenylamine 95%
Bis(4-formylphenyl)phenylamine

Precaución: for Research Use Only

Formula: C₂₀H₁₅NO₂
MW: 301.35 g/mol
MDL Number: MFCD03070375
CAS Number: 53566-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR