ω-Bromo-3-methoxyacetophenone 98%

Supplier: Ambeed
Danger

A341986-25G A341986-5G A341986-10G A341986-1G
76693-980EA 87 USD
76693-980 76693-982 77594-000 77626-650
ω-Bromo-3-methoxyacetophenone 98%
ω-Bromo-3-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD00000199
CAS Number: 5000-65-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR