4-((3-Hydroxypropyl)amino)-3-nitrophenol 95%

Supplier: Ambeed
Warning

A341372-100G A341372-25G A341372-5G A341372-1G
76693-970EA 99.73 USD
76693-970 76693-972 76693-974 77626-646
4-((3-Hydroxypropyl)amino)-3-nitrophenol 95%
4-((3-Hydroxypropyl)amino)-3-nitrophenol

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄
MW: 212.2000 g/mol
MDL Number: MFCD07368637
CAS Number: 92952-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR