Bromoacetyl chloride 98%

Supplier: Ambeed
Danger

A336964-1G A336964-25G A336964-5G
76693-948EA 6 USD
76693-948 77550-964 77550-966
Bromoacetyl chloride 98%
Bromoacetyl chloride

Precaución: for Research Use Only

Formula: BrCH₂COOCl
MW: 157.39 g/mol
MDL Number: MFCD00000724
CAS Number: 22118-09-8
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR