9,9-Diphenylfluorene 98%

Supplier: Ambeed
Warning

A335954-1G A335954-25G A335954-5G
76693-942EA 7.43 USD
76693-942 76693-944 76693-946
9,9-Diphenylfluorene 98%
9,9-Diphenylfluorene

Precaución: for Research Use Only

Formula: C₂₅H₁₈
MDL Number: MFCD00093944
CAS Number: 20302-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR