Chelerythrine chloride 99%

Supplier: Ambeed
Warning

A333732-100MG A333732-250MG
76693-928EA 6.37 USD
76693-928 77626-640 77626-642
Chelerythrine chloride 99%
Chelerythrine chloride

Precaución: for Research Use Only

Formula: C₂₁H₁₈ClNO₄
MDL Number: MFCD00060717
CAS Number: 3895-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR