1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-(((Benzyloxy)carbonyl)amino)cyclobutanecarboxylic acid

A333047-100MG A333047-1G A333047-250MG A333047-5G
76693-918EA 23 USD
76693-918 76693-920 76693-922 76693-924
1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid 95%
1-(Benzyloxycarbonylamino)cyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD03426163
CAS Number: 190004-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR