Chloranilic acid 95%

Supplier: Ambeed
Warning

Synonyms: Chloranilicacid

A332732-100G A332732-25G A332732-5G
76693-906EA 141.1 USD
76693-906 76693-908 76693-910
Chloranilic acid 95%
Chloranilic acid

Precaución: for Research Use Only

Formula: C₆H₂Cl₂O₄
MW: 208.98 g/mol
MDL Number: MFCD00001596
CAS Number: 87-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR