N,N,N',N'-Tetramethyl-p-phenylenediamine 95%

Supplier: Ambeed
Warning

Synonyms: N,N,N,N-Tetramethyl-1,4-phenylenediamine, N,N,N′,N′-Tetramethyl-p-phenylenediamine, TMPD, N,N,N',N'-Tetramethyl-1,4-phenylenediamine

A331721-10G A331721-1G A331721-25G A331721-5G
76693-900EA 24 USD
76693-900 77601-494 77626-634 77879-707
N,N,N',N'-Tetramethyl-p-phenylenediamine 95%
N,N,N',N'-Tetramethyl-p-phenylenediamine

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂
MDL Number: MFCD00008309
CAS Number: 100-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR