cis-6-Nonenal 98%

Supplier: Ambeed
Warning

A329667-5G A329667-1G A329667-25G
76693-876EA 59 USD
76693-876 77879-701 77879-702
cis-6-Nonenal 98%
cis-6-Nonenal

Precaución: for Research Use Only

Formula: C₉H₁₆O
MDL Number: MFCD00014683
CAS Number: 2277-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR